Product Details

SNP ID
rs348362
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:6466471 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CACCTGTCCCCTCTCTGCAGCGTCC[A/G]GAAGCCATCACTGCTATCATCGTGG
Phenotype
MIM: 609737 MIM: 613634 MIM: 608746
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRB3 PubMed Links

Gene Details

Gene
CRB3
Gene Name
crumbs 3, cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139161.4 381 Silent Mutation CCA,CCG P54P NP_631900.1
NM_174881.3 381 Silent Mutation CCA,CCG P54P NP_777377.1
NM_174882.2 381 Silent Mutation CCA,CCG P54P NP_777378.1
XM_017027471.1 381 Silent Mutation CCA,CCG P117P XP_016882960.1
Gene
DENND1C
Gene Name
DENN domain containing 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290331.1 381 Intron NP_001277260.1
NM_024898.3 381 Intron NP_079174.2
XM_006722905.2 381 Intron XP_006722968.2
XM_006722906.3 381 Intron XP_006722969.1
XM_011528318.2 381 Intron XP_011526620.1
Gene
SLC25A23
Gene Name
solute carrier family 25 member 23
There are no transcripts associated with this gene.

View Full Product Details