Product Details

SNP ID
rs2242604
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:73241639 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGGCTGAGAATTCTTCTCTCCCTGC[C/T]CTCTGTGAGTCTGCAGGGGGTGTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928123 PubMed Links

Gene Details

Gene
LOC101928123
Gene Name
uncharacterized LOC101928123
There are no transcripts associated with this gene.

Gene
PAPLN
Gene Name
papilin, proteoglycan like sulfated glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173462.3 Intron NP_775733.3
XM_011537290.2 Intron XP_011535592.1
XM_011537291.2 Intron XP_011535593.1
XM_011537292.2 Intron XP_011535594.1
XM_011537293.2 Intron XP_011535595.1
XM_011537294.2 Intron XP_011535596.1
XM_011537295.2 Intron XP_011535597.1
XM_011537296.2 Intron XP_011535598.1
XM_017021749.1 Intron XP_016877238.1
XM_017021750.1 Intron XP_016877239.1

View Full Product Details