Product Details
- SNP ID
-
rs734418
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31411182 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTGTCACAAAAAACTGTGAAACAAA[A/T]TGCCTTGATTGCTTCGTGAGCTAAA
- Phenotype
-
MIM: 616075
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB121
PubMed Links
Gene Details
- Gene
- DEFB121
- Gene Name
- defensin beta 121
View Full Product Details