Product Details

SNP ID
rs1728817
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10995440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTAGTTCCCGTGGGCCTGCCAT[G/T]GGAGCAGTGGCTGGTCCTCTACCTT
Phenotype
MIM: 137165
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
SLC6A1 PubMed Links

Gene Details

Gene
SLC6A1
Gene Name
solute carrier family 6 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003042.3 Intron NP_003033.3
XM_005265410.4 Intron XP_005265467.1
XM_005265411.4 Intron XP_005265468.1
XM_006713306.3 Intron XP_006713369.1
XM_011534025.2 Intron XP_011532327.1
XM_011534027.2 Intron XP_011532329.1
XM_017007071.1 Intron XP_016862560.1
XM_017007072.1 Intron XP_016862561.1

View Full Product Details