Product Details

SNP ID
rs4788460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72120610 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTCAAGAACTTGGGTGAGTTACT[C/T]ATCCCCATTTTACAGATTAGGAAAT
Phenotype
MIM: 605584
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHX38 PubMed Links

Gene Details

Gene
DHX38
Gene Name
DEAH-box helicase 38
There are no transcripts associated with this gene.

Gene
PMFBP1
Gene Name
polyamine modulated factor 1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160213.1 Intron NP_001153685.1
NM_031293.2 Intron NP_112583.2
XM_011523357.2 Intron XP_011521659.1
XM_011523358.2 Intron XP_011521660.1
XM_011523360.2 Intron XP_011521662.1
XM_011523361.2 Intron XP_011521663.1
XM_011523362.2 Intron XP_011521664.1

View Full Product Details