Product Details

SNP ID
rs574764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63974140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGATTCTGTAGAGGTTTTCTT[A/C]TTATTATTTATTTATTTACTTGTTT
Phenotype
MIM: 612086 MIM: 601697
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HMSD PubMed Links

Gene Details

Gene
HMSD
Gene Name
histocompatibility minor serpin domain containing
There are no transcripts associated with this gene.

Gene
SERPINB8
Gene Name
serpin family B member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031848.1 Intron NP_001027018.1
NM_001276490.1 Intron NP_001263419.1
NM_002640.3 Intron NP_002631.3
NM_198833.1 Intron NP_942130.1
XM_006722484.2 Intron XP_006722547.1
XM_011526025.1 Intron XP_011524327.1
XM_017025790.1 Intron XP_016881279.1
XM_017025791.1 Intron XP_016881280.1
XM_017025792.1 Intron XP_016881281.1

View Full Product Details