Product Details

SNP ID
rs684844
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:56931366 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTAGATAGATACATCAACCCAAA[A/G]TACAATGCACAAAGAGCTATAATAG
Phenotype
MIM: 120960
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C8B PubMed Links

Gene Details

Gene
C8B
Gene Name
complement component 8, beta polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000066.3 Intron NP_000057.2
NM_001278543.1 Intron NP_001265472.1
NM_001278544.1 Intron NP_001265473.1
XM_017002235.1 Intron XP_016857724.1

View Full Product Details