Product Details
- SNP ID
-
rs1054122
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52487215 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCTTTATTTAGCAATTGCAAACAG[C/T]GAAGAGCACAGAAATCATCACAGAG
- Phenotype
-
MIM: 148041
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT6A
PubMed Links
Gene Details
- Gene
- KRT6A
- Gene Name
- keratin 6A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005554.3 |
2409 |
UTR 3 |
|
|
NP_005545.1 |
View Full Product Details