Product Details

SNP ID
rs829864
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:98456417 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTATCTGAGAGCTGTCTCATAATC[C/T]TCATGAGCTTCACCATACCATACCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC9A7P1 PubMed Links

Gene Details

Gene
SLC9A7P1
Gene Name
solute carrier family 9 member 7 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details