Product Details

SNP ID
rs3822226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:13541050 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGAGCACTGGAGTCGCAGAGGGC[A/G]GAAGGTAGACACTGAGGGCCAGGCA
Phenotype
MIM: 602183
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01096 PubMed Links

Gene Details

Gene
LINC01096
Gene Name
long intergenic non-protein coding RNA 1096
There are no transcripts associated with this gene.

Gene
LINC01097
Gene Name
long intergenic non-protein coding RNA 1097
There are no transcripts associated with this gene.

Gene
NKX3-2
Gene Name
NK3 homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001189.3 2021 UTR 3 NP_001180.1

View Full Product Details