Product Details

SNP ID
rs11878687
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:41844991 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGGCGATGCTTGAAGTTCATTGGTC[C/T]TTGTGAGACAGGGTAAGAAAAGCAG
Phenotype
MIM: 614806
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMRTC2 PubMed Links

Gene Details

Gene
DMRTC2
Gene Name
DMRT like family C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040283.2 53 Intron NP_001035373.1
XM_011527214.1 53 Intron XP_011525516.1
XM_011527215.1 53 UTR 5 XP_011525517.1
XM_011527216.2 53 Intron XP_011525518.1
XM_017027123.1 53 Intron XP_016882612.1
XM_017027124.1 53 Intron XP_016882613.1
XM_017027125.1 53 Intron XP_016882614.1
XM_017027126.1 53 UTR 5 XP_016882615.1
XM_017027127.1 53 Intron XP_016882616.1
XM_017027128.1 53 UTR 5 XP_016882617.1
XM_017027129.1 53 Intron XP_016882618.1
Gene
LYPD4
Gene Name
LY6/PLAUR domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291419.1 53 Intron NP_001278348.1
NM_173506.6 53 Intron NP_775777.3
XM_005258543.3 53 Intron XP_005258600.1
XM_005278383.4 53 Intron XP_005278440.1
XM_005278385.4 53 Intron XP_005278442.1
XM_005278387.4 53 Intron XP_005278444.1
XM_005278388.3 53 Intron XP_005278445.1
XM_006723035.2 53 Intron XP_006723098.1
XM_011526483.2 53 Intron XP_011524785.1
XM_011526484.2 53 Intron XP_011524786.1

View Full Product Details