Product Details

SNP ID
rs7259215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41712675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGAGTCACTGATTTCAAACCAC[C/T]TTCAGCATCTTCCTTTGAGCATAAT
Phenotype
MIM: 114890
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEACAM5 PubMed Links

Gene Details

Gene
CEACAM5
Gene Name
carcinoembryonic antigen related cell adhesion molecule 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291484.2 Intron NP_001278413.1
NM_001308398.1 Intron NP_001295327.1
NM_004363.5 Intron NP_004354.3
XM_005258413.3 Intron XP_005258470.2
XM_011526322.1 Intron XP_011524624.1
XM_017026145.1 Intron XP_016881634.1
XM_017026146.1 Intron XP_016881635.1

View Full Product Details