Product Details

SNP ID
rs906594
Assay Type
Functionally tested
NCBI dbSNP Submissions
62
Location
Chr.1:154945225 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGAAGCTGACTTGAGCAGGCACC[A/G]CCAAGCATATGTGGGAAAAACCTGG
Phenotype
MIM: 607622
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PBXIP1 PubMed Links

Gene Details

Gene
PBXIP1
Gene Name
PBX homeobox interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317734.1 Intron NP_001304663.1
NM_001317735.1 Intron NP_001304664.1
NM_020524.3 Intron NP_065385.2
Gene
PMVK
Gene Name
phosphomevalonate kinase
There are no transcripts associated with this gene.

View Full Product Details