Product Details

SNP ID
rs2306434
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:207825181 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAAGTACTTTGTGGCCTTCCTGCCG[A/C]GGTGCAAAATGAGGTCTATTTTAGG
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
PLEKHM3 PubMed Links

Gene Details

Gene
PLEKHM3
Gene Name
pleckstrin homology domain containing M3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080475.2 5914 UTR 3 NP_001073944.1
XM_005246545.3 5914 UTR 3 XP_005246602.1
XM_006712521.3 5914 Intron XP_006712584.1
XM_011511160.2 5914 UTR 3 XP_011509462.1
XM_011511163.2 5914 Intron XP_011509465.1
XM_017004071.1 5914 Intron XP_016859560.1
XM_017004072.1 5914 UTR 3 XP_016859561.1
XM_017004073.1 5914 UTR 3 XP_016859562.1

View Full Product Details