Product Details

SNP ID
rs11809891
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:160541777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCAGATTGCTAAGGAGTGTGGTT[C/G]CCCAGGAAAAACATTTTAGTGTGGA
Phenotype
MIM: 604513
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD84 PubMed Links

Gene Details

Gene
CD84
Gene Name
CD84 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184879.1 7235 UTR 3 NP_001171808.1
NM_001184881.1 7235 UTR 3 NP_001171810.1
NM_001184882.1 7235 UTR 3 NP_001171811.1
NM_003874.3 7235 UTR 3 NP_003865.1
XM_011510094.2 7235 UTR 3 XP_011508396.1
XM_011510095.2 7235 UTR 3 XP_011508397.1

View Full Product Details