Product Details

SNP ID
rs1677133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21637565 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGAACAAAATATATCTCAATTGT[A/G]AGGTCCCACTTGCCACTGTCACCTA
Phenotype
MIM: 150100
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LDHB PubMed Links

Gene Details

Gene
LDHB
Gene Name
lactate dehydrogenase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174097.2 Intron NP_001167568.1
NM_001315537.1 Intron NP_001302466.1
NM_002300.7 Intron NP_002291.1
XM_006719074.2 Intron XP_006719137.1

View Full Product Details