Product Details

SNP ID
rs198781
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:38801836 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGCAAGATTTTGGCATTTAGTAA[C/G]TTAAGTCTGAATACCGAGTTTTCAT
Phenotype
MIM: 300961
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MID1IP1 PubMed Links

Gene Details

Gene
MID1IP1
Gene Name
MID1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098790.1 Intron NP_001092260.1
NM_001098791.1 Intron NP_001092261.1
NM_021242.5 Intron NP_067065.1
Gene
MID1IP1-AS1
Gene Name
MID1IP1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details