Product Details

SNP ID
rs589034
Assay Type
Validated
NCBI dbSNP Submissions
63
Location
Chr.1:32224256 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CTCCTACTCCCTGGTCTCTTCACTG[A/G]CTGAGGAAACATTCAGGACAAATTC
Phenotype
MIM: 603911
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCDC2B PubMed Links

Gene Details

Gene
DCDC2B
Gene Name
doublecortin domain containing 2B
There are no transcripts associated with this gene.

Gene
EIF3I
Gene Name
eukaryotic translation initiation factor 3 subunit I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003757.3 Intron NP_003748.1
XM_017002671.1 Intron XP_016858160.1
Gene
MTMR9LP
Gene Name
myotubularin related protein 9-like, pseudogene
There are no transcripts associated with this gene.

Gene
TMEM234
Gene Name
transmembrane protein 234
There are no transcripts associated with this gene.

View Full Product Details