Product Details

SNP ID
rs34670199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:54146645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGATCTCTTTTCTCCCACAATA[C/T]GGGACATCCTCATCACTTTACCTTT
Phenotype
MIM: 126065
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP24A1 PubMed Links

Gene Details

Gene
CYP24A1
Gene Name
cytochrome P450 family 24 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000782.4 Intron NP_000773.2
NM_001128915.1 Intron NP_001122387.1
XM_005260304.4 Intron XP_005260361.1
XM_017027691.1 Intron XP_016883180.1
XM_017027692.1 Intron XP_016883181.1
XM_017027693.1 Intron XP_016883182.1

View Full Product Details