Product Details

SNP ID
rs764282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143912144 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAACTGAACATATAATATAATA[G/T]GTATTAGATGATGAGTTCTATAGAT
Phenotype
MIM: 608497 MIM: 616251
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2F1 PubMed Links

Gene Details

Gene
OR2F1
Gene Name
olfactory receptor family 2 subfamily F member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012369.2 Intron NP_036501.2
XM_011516019.2 Intron XP_011514321.1
XM_011516020.2 Intron XP_011514322.1
Gene
TCAF1
Gene Name
TRPM8 channel associated factor 1
There are no transcripts associated with this gene.

View Full Product Details