Product Details
- SNP ID
-
rs261247
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:65566375 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAATGATTATTAAGATTACAGCAGT[C/T]ATATCCCAGTTCCAAGCTATAGGAT
- Phenotype
-
MIM: 611502
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CENPK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114802254] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CENPK
- Gene Name
- centromere protein K
There are no transcripts associated with this gene.
- Gene
- PPWD1
- Gene Name
- peptidylprolyl isomerase domain and WD repeat containing 1
View Full Product Details