Product Details

SNP ID
rs456666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:77694131 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAAAAGTTATTGAGTTATTGATC[A/T]CATCGTGTGCACACTGCAGGAGGTT
Phenotype
MIM: 610058
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TBCA PubMed Links

Gene Details

Gene
TBCA
Gene Name
tubulin folding cofactor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297738.1 Intron NP_001284667.1
NM_001297740.1 Intron NP_001284669.1
NM_004607.2 Intron NP_004598.1

View Full Product Details