Product Details

SNP ID
rs10451452
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:55594855 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCCAGTTATCATGAGCTGAATACAG[C/G]CCTTAGGTGTGTTAGGGCAGAGAAG
Phenotype
MIM: 609133
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FIZ1 PubMed Links

Gene Details

Gene
FIZ1
Gene Name
FLT3 interacting zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032836.2 Intron NP_116225.2
XM_005259352.4 Intron XP_005259409.1
XM_011527426.2 Intron XP_011525728.1
Gene
ZNF524
Gene Name
zinc finger protein 524
There are no transcripts associated with this gene.

View Full Product Details