Product Details

SNP ID
rs4620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:21146817 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAAATCCAGGATGGATGCAAC[C/T]TTGAAAGAACTGACAAGCTTAGTAA
Phenotype
MIM: 602949
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAP18 PubMed Links

Gene Details

Gene
SAP18
Gene Name
Sin3A associated protein 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005870.4 291 Silent Mutation ACC,ACT T84T NP_005861.2
XM_011534868.2 291 Silent Mutation ACC,ACT T16T XP_011533170.1
Gene
SKA3
Gene Name
spindle and kinetochore associated complex subunit 3
There are no transcripts associated with this gene.

View Full Product Details