Product Details

SNP ID
rs11840534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:30934287 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATGGCTACTCTGTGCCACTTCAT[C/T]CCCCACTGCTGGGAGAGTCACCCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEDAG PubMed Links

Gene Details

Gene
MEDAG
Gene Name
mesenteric estrogen dependent adipogenesis
There are no transcripts associated with this gene.

Gene
TEX26
Gene Name
testis expressed 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152325.1 Intron NP_689538.1
XM_011534919.2 Intron XP_011533221.1
XM_011534921.2 Intron XP_011533223.1
XM_011534924.2 Intron XP_011533226.1
XM_011534925.2 Intron XP_011533227.1
XM_011534926.2 Intron XP_011533228.1
XM_011534927.2 Intron XP_011533229.1
XM_011534928.2 Intron XP_011533230.1
Gene
TEX26-AS1
Gene Name
TEX26 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details