Product Details

SNP ID
rs6667486
Assay Type
Validated
NCBI dbSNP Submissions
43
Location
Chr.1:20167377 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCATCAGCTCTCCCCCCATCTTCAC[A/G]AGCTCAGTATTTGGTGCCACTATCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLA2G2C PubMed Links

Gene Details

Gene
PLA2G2C
Gene Name
phospholipase A2 group IIC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316722.1 Intron NP_001303651.1
XM_017001270.1 Intron XP_016856759.1

View Full Product Details