Product Details

SNP ID
rs37810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3401881 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGGCTTTGTCTCTGCATCCCGTT[C/T]CCCCTAACATCCTCAGAGAACCTTC
Phenotype
MIM: 603900
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF174 PubMed Links

Gene Details

Gene
ZNF174
Gene Name
zinc finger protein 174
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032292.2 692 UTR 5 NP_001027463.1
NM_003450.2 692 UTR 5 NP_003441.1
Gene
ZSCAN32
Gene Name
zinc finger and SCAN domain containing 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284527.1 692 Intron NP_001271456.1
NM_001284528.1 692 Intron NP_001271457.1
NM_001284529.1 692 Intron NP_001271458.1
NM_001324340.1 692 Intron NP_001311269.1
NM_001324341.1 692 Intron NP_001311270.1
NM_001324342.1 692 Intron NP_001311271.1
NM_001324343.1 692 Intron NP_001311272.1
NM_001324344.1 692 Intron NP_001311273.1
NM_001324345.1 692 Intron NP_001311274.1
NM_001324346.1 692 Intron NP_001311275.1
NM_017810.3 692 Intron NP_060280.2
XM_005255403.2 692 Intron XP_005255460.1
XM_011522555.2 692 Intron XP_011520857.1
XM_017023371.1 692 Intron XP_016878860.1
XM_017023372.1 692 Intron XP_016878861.1
XM_017023373.1 692 Intron XP_016878862.1

View Full Product Details