Product Details

SNP ID
rs4151693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141150130 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGAGATAGATGTGTCCGGGAAGG[C/T]AGTCGTCGCCAGACAAGTTGTAAGA
Phenotype
MIM: 606332
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDHB17P PubMed Links

Gene Details

Gene
PCDHB17P
Gene Name
protocadherin beta 17 pseudogene
There are no transcripts associated with this gene.

Gene
PCDHB6
Gene Name
protocadherin beta 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303145.1 109 UTR 5 NP_001290074.1
NM_018939.3 109 UTR 5 NP_061762.2

View Full Product Details