Product Details

SNP ID
rs12976785
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48556533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGTCAGGAGAGGGCCAGCTGCC[A/G]TGGAAACTCCCCGTGGTGGGCCATC
Phenotype
MIM: 604125
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SULT2B1 PubMed Links

Gene Details

Gene
SULT2B1
Gene Name
sulfotransferase family 2B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004605.2 Intron NP_004596.2
NM_177973.1 Intron NP_814444.1

View Full Product Details