Product Details

SNP ID
rs1811167
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:6071980 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGCATGGGATCTGAAGGAGGCTTGG[C/T]GGTGGTTGGGCTGGTGGCTGAGCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WSCD1 PubMed Links

Gene Details

Gene
WSCD1
Gene Name
WSC domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015253.1 Intron NP_056068.1
XM_005256572.1 Intron XP_005256629.1
XM_005256573.1 Intron XP_005256630.1
XM_011523777.1 Intron XP_011522079.1

View Full Product Details