Product Details

SNP ID
rs35585009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:38942254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCTGAGGCTTTTAGGAAATCC[A/C]CAAATATGAATATATATAGTATGTA
Phenotype
MIM: 601743 MIM: 609022
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OSMR PubMed Links

Gene Details

Gene
OSMR
Gene Name
oncostatin M receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168355.2 5092 Intron NP_001161827.1
NM_001323504.1 5092 Intron NP_001310433.1
NM_001323505.1 5092 Intron NP_001310434.1
NM_001323506.1 5092 Intron NP_001310435.1
NM_001323507.1 5092 Intron NP_001310436.1
NM_003999.2 5092 Intron NP_003990.1
XM_005248384.1 5092 Intron XP_005248441.1
XM_005248386.2 5092 Intron XP_005248443.1
XM_005248387.2 5092 Intron XP_005248444.1
XM_011514161.2 5092 Intron XP_011512463.1
XM_017010019.1 5092 Intron XP_016865508.1
XM_017010020.1 5092 Intron XP_016865509.1
Gene
RICTOR
Gene Name
RPTOR independent companion of MTOR complex 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285439.1 5092 UTR 3 NP_001272368.1
NM_001285440.1 5092 UTR 3 NP_001272369.1
NM_152756.4 5092 UTR 3 NP_689969.2
XM_006714463.3 5092 UTR 3 XP_006714526.1
XM_011514005.2 5092 UTR 3 XP_011512307.1
XM_011514006.2 5092 UTR 3 XP_011512308.1
XM_017009311.1 5092 UTR 3 XP_016864800.1
XM_017009312.1 5092 UTR 3 XP_016864801.1
XM_017009313.1 5092 UTR 3 XP_016864802.1
XM_017009314.1 5092 UTR 3 XP_016864803.1
XM_017009315.1 5092 UTR 3 XP_016864804.1

View Full Product Details