Product Details

SNP ID
rs62191702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:229943057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACTCCCCAATCCAAGCCAATGA[A/G]ATGCTTAAGAAAGGCGCACTTCCTT
Phenotype
MIM: 609105
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO36 PubMed Links

Gene Details

Gene
FBXO36
Gene Name
F-box protein 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174899.4 Intron NP_777559.3
XM_005246317.1 Intron XP_005246374.1

View Full Product Details