Product Details

SNP ID
rs6037941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:4735833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGCTCTTCCTTTGACACACAGAA[A/G]ATAGCTTTTTATCTCACTGGGATCT
Phenotype
MIM: 604263
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRND PubMed Links

Gene Details

Gene
PRND
Gene Name
prion protein 2 (dublet)
There are no transcripts associated with this gene.

Gene
PRNT
Gene Name
prion protein (testis specific)
There are no transcripts associated with this gene.

View Full Product Details