Product Details

SNP ID
rs175436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75141980 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCATTGAGGCTTTATATCACAGTA[C/G]AAAAAGCTAAAAAATCCTCAATATC
Phenotype
MIM: 605406
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMED10 PubMed Links

Gene Details

Gene
TMED10
Gene Name
transmembrane p24 trafficking protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006827.5 Intron NP_006818.3

View Full Product Details