Product Details

SNP ID
rs2071417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6917143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGGAGAAAGTGGGGAAGCGTCA[C/G]GGTGGGGAGGCGTGGAGCAGATAGA
Phenotype
MIM: 607462 MIM: 131360
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATN1 PubMed Links

Gene Details

Gene
ATN1
Gene Name
atrophin 1
There are no transcripts associated with this gene.

Gene
ENO2
Gene Name
enolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001975.2 Intron NP_001966.1
Gene
LRRC23
Gene Name
leucine rich repeat containing 23
There are no transcripts associated with this gene.

View Full Product Details