Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001010855.3 | 2227 | UTR 3 | NP_001010855.1 | ||
NM_001290211.1 | 2227 | UTR 3 | NP_001277140.1 | ||
XM_011523677.2 | 2227 | Silent Mutation | ACA,ACG | T762T | XP_011521979.1 |
XM_011523678.2 | 2227 | Silent Mutation | ACA,ACG | T758T | XP_011521980.1 |
XM_011523680.2 | 2227 | Silent Mutation | ACA,ACG | T722T | XP_011521982.1 |
XM_011523681.2 | 2227 | Silent Mutation | ACA,ACG | T683T | XP_011521983.1 |
XM_011523683.2 | 2227 | Silent Mutation | ACA,ACG | T683T | XP_011521985.1 |
XM_011523686.2 | 2227 | Silent Mutation | ACA,ACG | T474T | XP_011521988.1 |
XM_017024228.1 | 2227 | Silent Mutation | ACA,ACG | T763T | XP_016879717.1 |
XM_017024229.1 | 2227 | UTR 3 | XP_016879718.1 | ||
XM_017024230.1 | 2227 | UTR 3 | XP_016879719.1 | ||
XM_017024231.1 | 2227 | Silent Mutation | ACA,ACG | T683T | XP_016879720.1 |