Product Details

SNP ID
rs2920282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142676345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCACAGTGGAGGACGCTTAGTCA[C/T]GGGGCATACCAGGCACATGGTGGGA
Phenotype
MIM: 603210 MIM: 602470
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JRK PubMed Links

Gene Details

Gene
JRK
Gene Name
Jrk helix-turn-helix protein
There are no transcripts associated with this gene.

Gene
PSCA
Gene Name
prostate stem cell antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005672.4 Intron NP_005663.2

View Full Product Details