Product Details

SNP ID
rs916791
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18613347 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGATGCAGGTCCTTGCGATAAGG[A/G]GCCGATACCACCTCCCGGGCTCCTT
Phenotype
MIM: 604237
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CRLF1 PubMed Links

Gene Details

Gene
CRLF1
Gene Name
cytokine receptor like factor 1
There are no transcripts associated with this gene.

Gene
TMEM59L
Gene Name
transmembrane protein 59 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012109.2 Intron NP_036241.1

View Full Product Details