Product Details

SNP ID
rs1207577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:24363270 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGGCATATTCTTCTACGTCCAAC[C/G]ATGCAGACTTTAGCCCATGATGTGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SUPT20HL1 PubMed Links

Gene Details

Gene
SUPT20HL1
Gene Name
SPT20 homolog, SAGA complex component-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136234.1 510 Silent Mutation ACC,ACG T170T NP_001129706.1

View Full Product Details