Product Details

SNP ID
hCV7428637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:235112102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCAGCCAAGCTCTGAGCACTTA[C/G]AATTCTCTGAAAGAAAAAAAAAATA
Phenotype
MIM: 601848
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TOMM20 PubMed Links

Gene Details

Gene
TOMM20
Gene Name
translocase of outer mitochondrial membrane 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014765.2 Intron NP_055580.1

View Full Product Details