Product Details

SNP ID
rs1045905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40019241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTAGAAAGTTCCTCAGAGGTAA[A/G]ACTACTCCTGGGCTGGGGCGGGCGC
Phenotype
MIM: 138970 MIM: 607000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSF3 PubMed Links

Gene Details

Gene
CSF3
Gene Name
colony stimulating factor 3
There are no transcripts associated with this gene.

Gene
MED24
Gene Name
mediator complex subunit 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079518.1 3441 UTR 3 NP_001072986.1
NM_001267797.1 3441 UTR 3 NP_001254726.1
NM_014815.3 3441 UTR 3 NP_055630.2
XM_005257874.2 3441 UTR 3 XP_005257931.1
XM_006722204.2 3441 UTR 3 XP_006722267.1
XM_006722206.2 3441 UTR 3 XP_006722269.1
XM_006722207.2 3441 UTR 3 XP_006722270.1
XM_011525529.2 3441 UTR 3 XP_011523831.1
XM_011525530.2 3441 UTR 3 XP_011523832.1
XM_011525531.2 3441 UTR 3 XP_011523833.1
XM_011525532.2 3441 UTR 3 XP_011523834.1
XM_011525533.2 3441 UTR 3 XP_011523835.1
XM_011525534.2 3441 UTR 3 XP_011523836.1
XM_011525535.2 3441 UTR 3 XP_011523837.1
XM_011525536.2 3441 UTR 3 XP_011523838.1
XM_011525538.2 3441 UTR 3 XP_011523840.1
XM_017025458.1 3441 UTR 3 XP_016880947.1
XM_017025459.1 3441 UTR 3 XP_016880948.1
XM_017025460.1 3441 UTR 3 XP_016880949.1
XM_017025461.1 3441 UTR 3 XP_016880950.1
XM_017025462.1 3441 UTR 3 XP_016880951.1
XM_017025463.1 3441 UTR 3 XP_016880952.1
XM_017025464.1 3441 UTR 3 XP_016880953.1
XM_017025465.1 3441 UTR 3 XP_016880954.1
XM_017025466.1 3441 UTR 3 XP_016880955.1
XM_017025467.1 3441 UTR 3 XP_016880956.1
XM_017025468.1 3441 UTR 3 XP_016880957.1
XM_017025469.1 3441 UTR 3 XP_016880958.1
XM_017025470.1 3441 UTR 3 XP_016880959.1
XM_017025471.1 3441 Intron XP_016880960.1
Gene
MIR6884
Gene Name
microRNA 6884
There are no transcripts associated with this gene.

Gene
SNORD124
Gene Name
small nucleolar RNA, C/D box 124
There are no transcripts associated with this gene.

View Full Product Details