Product Details

SNP ID
rs868192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:49561042 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGACGCAGACTCCGGGCCAAGTT[C/T]TCCCTCCGCTCGCTGCTTTCTGCCG
Phenotype
MIM: 603684
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIPG PubMed Links

Gene Details

Gene
LIPG
Gene Name
lipase G, endothelial type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308006.1 344 Intron NP_001294935.1
NM_006033.3 344 Intron NP_006024.1
XM_005258390.1 344 UTR 5 XP_005258447.1
XM_011526265.2 344 Intron XP_011524567.1
XM_011526267.1 344 UTR 5 XP_011524569.1
XM_017026095.1 344 UTR 5 XP_016881584.1

View Full Product Details