Product Details

SNP ID
rs1789514
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.18:36112691 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAATGGTATTTTGGCTGTGAATGAA[C/T]AGAAGTTTCCCAAACCAACTCTGTA
Phenotype
MIM: 608731
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC39A6 PubMed Links

Gene Details

Gene
SLC39A6
Gene Name
solute carrier family 39 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099406.1 Intron NP_001092876.1
NM_012319.3 Intron NP_036451.3
XM_011525900.1 Intron XP_011524202.1
XM_011525901.1 Intron XP_011524203.1

View Full Product Details