Product Details
- SNP ID
-
rs699463
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:24544675 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CAGCAACTAAAGTGCTTGTTAGCCT[A/G]TGAGCCCTACAAACACCCACTCCCA
- Phenotype
-
MIM: 610045
MIM: 609269
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALDH5A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2760183] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALDH5A1
- Gene Name
- aldehyde dehydrogenase 5 family member A1
There are no transcripts associated with this gene.
- Gene
- KIAA0319
- Gene Name
- KIAA0319
View Full Product Details