Product Details

SNP ID
rs1459628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:19267242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAGACACAGATTTTAAAATACA[A/G]CTCAGAGGAAATGCATATGTAAAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD26P3 PubMed Links

Gene Details

Gene
ANKRD26P3
Gene Name
ankyrin repeat domain 26 pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details