Product Details
- SNP ID
-
hCV7472567
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:121410916 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGCAACATTTTTGCCTGGGACACT[C/T]TGGAGGTTGGCTTGGGCTTTTTTGT
- Phenotype
-
MIM: 609078
MIM: 608299
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KDM2B
PubMed Links
Gene Details
- Gene
- KDM2B
- Gene Name
- lysine demethylase 2B
There are no transcripts associated with this gene.
- Gene
- RNF34
- Gene Name
- ring finger protein 34
There are no transcripts associated with this gene.
View Full Product Details