Product Details

SNP ID
rs1043604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:33013273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCCCCTACCCCAGATAAAAAGA[A/T]TCAGACTGTACAAGCTGTGTCACAG
Phenotype
MIM: 601784
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ASIC2 PubMed Links

Gene Details

Gene
ASIC2
Gene Name
acid sensing ion channel subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001094.4 3257 UTR 3 NP_001085.2
NM_183377.1 3257 UTR 3 NP_899233.1

View Full Product Details