Product Details

SNP ID
rs1804538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40392615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTCCGTGGCTCTGTTTCTCGTG[A/G]AGGGACATCAAAATTACTTTCGTCA
Phenotype
MIM: 126430
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TOP2A PubMed Links

Gene Details

Gene
TOP2A
Gene Name
topoisomerase (DNA) II alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001067.3 4035 Missense Mutation CCA,TCA P1312S NP_001058.2
XM_005257632.1 4035 Missense Mutation CCA,TCA P1300S XP_005257689.1
XM_011525165.1 4035 Missense Mutation CCA,TCA P1312S XP_011523467.1

View Full Product Details