Product Details

SNP ID
rs1050588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27598158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCTGGTTACTTGGTGACGTGCA[A/G]AGCTCTCTCTGGGGGGCTGCAGCTC
Phenotype
MIM: 185430 MIM: 132811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLU PubMed Links

Gene Details

Gene
CLU
Gene Name
clusterin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001831.3 3525 UTR 3 NP_001822.3
XM_006716284.2 3525 UTR 3 XP_006716347.1
Gene
EPHX2
Gene Name
epoxide hydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256482.1 3525 Intron NP_001243411.1
NM_001256483.1 3525 Intron NP_001243412.1
NM_001256484.1 3525 Intron NP_001243413.1
NM_001979.5 3525 Intron NP_001970.2
XM_017013199.1 3525 Intron XP_016868688.1
XM_017013200.1 3525 Intron XP_016868689.1

View Full Product Details