Product Details

SNP ID
rs826953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:43678452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCCAGGTTAGCTTGGTTTTTGG[A/T]TTGTTTTTTCTAACTCTGAACTGGA
Phenotype
MIM: 601415
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MYBL2 PubMed Links

Gene Details

Gene
MYBL2
Gene Name
MYB proto-oncogene like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278610.1 Intron NP_001265539.1
NM_002466.3 Intron NP_002457.1

View Full Product Details